VWR® Probe One-Step RT-qPCR Kit
Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
73222-0500EA 4830 DKK
73222-0500 73222-0100 73222-1000 73222-2500
VWR® Probe One-Step RT-qPCR Kit
Nukleinsyrereagenser qPCR/RT-qPCR Enzymes and Kits
VWR® Probe One-Step RT-qPCR Kit is designed to facilitate rapid, sensitive, and precise detection and quantification of various RNA sequences via hydrolysis probes. It features a distinctive universal passive reference dye that is compatible with ROX-dependent and ROX-independent qPCR instruments.
  • For the detection and quantification of RNA targets using probe-based technology, such as TaqMan®
  • Ready-to-use kit containing all necessary reaction components, except for primers, probes, and RNA template
  • Suitable for a variety of RNA sample types
  • Includes Hot Start Taq DNA Polymerase with VWR® Reverse Transcriptase for improved reaction specificity and room temperature setup
  • For multiplex applications of up to 5 targets
  • Incorporates dUTP for carryover contamination prevention
  • VWR® Probe One-Step Reaction Mix is supplied at a 2X concentration
The VWR® RT Enzyme Mix, supplied at a 20X concentration, contains the specially designed, temperature-controlled VWR® Reverse Transcriptase and Murine RNase Inhibitor to assist in preventing RNA degradation. This temperature-controlled RT enzyme prevents unwanted non-specific activity and displays superior thermostability in comparison to other reverse transcriptases.
Advarsel: Store at -20°C. For research use only.
Bestil nu
Specifikationer
Environmentally Preferable
Ansøgning RT-qPCR probe
One step Yes