Geldokumentationssystemer, PhotoDoc-It™ 70 Imaging System
Leverandør: UVP ULTRA VIOLET PRODUCTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
97-0274-05 97-0274-02 97-0274-08 97-0274-11 97-0274-14
732-1249EA 64430 DKK
732-1249 732-1248 732-1226 732-1227 732-1228
Geldokumentationssystemer, PhotoDoc-It™ 70 Imaging System
Fotodokumentationssystemer
PhotoDoc-It™ 70 er designet til at tage, udskrive og gemme farvebilleder til grundlæggende geldokumentation uden behov for film.
  • DigiCam 70 colour camera with >16 megapixel resolution and 5,0× optical zoom
  • Lightweight hood provides a compact darkroom environment
  • Save images to memory card or instantly generate 10×15 cm (4×6") low cost colour prints using the photo printer included
  • Ethidium bromide filter for fluorescent stained gels (EtBr Red 570 to 640 nm), positioned inside hood, is easily interchangeable for other filter types
  • Combination lock