Pipette tips, low retention, extended length, ART™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3510-05 3151-05-HR 3152-05-HR 3511-05-HR 3512-05-HR 3541-05-HR 3542-05-HR 3791-05-HR 3792-05-HR
732-0321EA 166 DKK
732-0321 613-2454 613-2456 613-2466 613-2468 613-2478 613-2487 613-2781 613-2783
Pipette tips, low retention, extended length, ART™
Pipettespidser
Using advanced low-retention technology, the hydrophobic inner surface of these tips significantly reduces sample loss to adhesion and maximises sample recovery.
  • Non sterile tips are certified to be free of RNase, DNase, DNA, ATP, endotoxins, pyrogens and PCR inhibitors
  • Sterile tips are certified to be free of RNase, DNase, DNA, ATP, bioburden, endotoxins, pyrogens and PCR inhibitors
  • Extended length to prevent contamination by keeping the pipettete shaft from entering the sample vessel
  • Rack cover can easily be realigned from a traditional lift-off to a