Dichlormethan, BAKER, Laboratory Reagent, J.T.Baker®

Leverandør: Avantor
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Methylenchlorid

7316.2500EA 723 DKK
7316.2500
Dichlormethan, BAKER, Laboratory Reagent, J.T.Baker®
Dichloromethane
Formel: CH₂Cl₂
Molekylvægt: 84.93 g/mol
Kogepunkt: 39.8
Smeltepunkt: –95 °C
Massefylde: 1.322 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000881
CAS-nummer: 75-09-2
EINECS: 200-838-9
UN nummer: 1593
ADR: 6.1 III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Assay (by GC) min. 99 %
Boiling Range 38 - 41 °C
Preservative (as Amylene) about 20 ppm
Residue after Evaporation max. 0.005 %
Water (H2O) max. 0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR