PCR plate, 96-well, automation compatible, Axygen®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PCR-96-HS-AC-C
731-0678EA 360 DKK
731-0678
PCR plate, 96-well, automation compatible, Axygen®
Mikroplader PCR plader
This automation-compatible, polypropylene PCR microplate features a rigid frame with a small split that sustains heat transferring during and post thermal cycling without distorting its shape. The plate also features raised rims around the wells to provide tight sealing to minimise heat evaporation.
  • Remains flat during and post thermal cycling
  • Raised well rims help reduce heat evaporation
  • Clear wells, visible grid for easy sample loading and tracking
  • Ultra-thin and consistent wall thickness for easy and consistent heat transfer
  • Single notch corner
  • SBS