Automated tissue processor, MTM
Leverandør: Slee
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
11000500
720-1974EA 394440 DKK
720-1974
Automated tissue processor, MTM
Vævsbehandlere
The MTM II fast tissue processor offers a cost-efficient and user-friendly alternative for fast biopsies processing and overnight batch tissue processing needs of every histology laboratory. The instrument can either be used for fully automated fast biopsy processing or high volume overnight batches of up to 300 samples.
  • Safe and homogeneous tissue processing with preheated reagents (EHE)
  • Fully automatic reagent management system
  • Automatic cleaning cycles
  • Intuitive control via 15'' touchscreen display
  • Charcoal filter supported ventilation
  • Remote alarm