Colour standards, starter set for Lico Spectral colorimeter
Leverandør: Hach
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
LZM354
705-1122EA 1990 DKK
705-1122
Colour standards, starter set for Lico Spectral colorimeter
Standarder Farve Referenser og Spektroskopi Standarder
Calibration solutions for Lico spectral colorimeter.
Leveringsinformation: Starter Kit consists of 6×ADDISTA certified colour standard colour solutions, 10× glass round cuvettes, 11 mm, and 10×PMMA rectangular cuvettes, 50 mm