Synlige spektrofotometre, DR3900

Leverandør: Hach
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

LPV4409900001 LPV4409800001 LPV4409910001
705-0764EA 59970 DKK
705-0764 705-1062 705-1147
Synlige spektrofotometre, DR3900
Spektrofotometer og fotometer UV/Vis-spektrofotometre
Visible spectrophotometer to ensure the reliability and traceability of measurement results during routine analysis.

  • Integrated barcode reader (IBR+) increases the reliability of measurement values
  • Data updates and quality assurance with AQA+
  • Alignment of laboratory and process analysis by LINK2SC
  • Key methods are possible including COD, chlorine, nitrates and ammonia
  • High quality reagents formulated to ensure outstanding performance and results
  • Transmittance (%) - absorbance - concentration - scanning

Radio frequency identification (RFID) guaranteesMore Product Information
Bestil nu

Specifikationer

Environmentally Preferable
Display 7" LCD
Antal metoder Over 220 (pre-programmable) and 100 (user-defined)
Optisk system Spectral reference beam
Absorbansområde −3 - +3 A
Absorbansnøjagtighed ±5 mA (0 - 0,5 A); 1% (0,5 - 2,0 A)
Wavelength range 320 - 1100 nm
Kuvetter Rectangular: 10, 20, 30, 50 mm, 1" − Round: Ø 13 mm, 1"
Elektriske specifikationer 110 - 240 V
Interface USB type A (2); USB type B; Ethernet
Lyskilde Tungsten
Hukommelse 2000 values including result, date, time, sample ID, operator ID
Spektral båndbredde 5 nm
Straylight Less than 0,1% T @ 340 nm with NaNO₂
W×D×H 255×350×151 mm
Vægt 4,2 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR