UV/Visible spectrophotometer, photoLab® 7600
Leverandør: WTW
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
250204
705-0922EA 83690 DKK
705-0922
UV/Visible spectrophotometer, photoLab® 7600
Spektrofotometer og fotometer UV/Vis-spektrofotometre
The pHotoLab® 7600 combines routine analysis with spectral analysis and pioneering procedure OptRF.
  • Pioneering measurement procedure 'OptRF', reagent-free for COD, nitrate and nitrite in the UV range
  • Brilliant colour screen operation guidance
  • Multi-step and multi-wavelength programming tools
  • photoLab® color+data management included
  • Routine analysis with barcode recognition and automatic measurement range adaption for all cuvette sizes
The important parameters COD, nitrate and nitrite of municipal waste water samples from the outlet can be measuredMore Product Information