CultiControl freeze-dried microorganism strains derived from NCTC® cultures
Leverandør: LIOFILCHEM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
70174 70114 70169 70119 70021 70022 70155 70023 70113 70078 70111 70139 70147 70167 70145 70086 70024 70072 70159 70020 70090 70112 70053 70059 70032 70095 70138 70156 70026 70173 70115 70025 70171 70187 70163 70027 70028 70157 70099 70203 70077 70076 70120 70160 70176 70136 70142 70070 70192 70089 70080 70209 70082 70151 70052 70101 70029 70030 70188 70085 70060 70031 70148 70102 70096 70103 70104 70074 70164 70189 70165 70094 70162 70134 70190 70083 70108 70033 70034 70109 70110 70035 70036 70154 70132 70084 70037 70054 70121 70044 70039 70137 70041 70040 70043 70182 70202 70045 70126 70153 70133 70149 70046 70127 70061 70062 70047 70048 70186 70064 70144 70056 70168 70050
70174.EA 858 DKK
70174. 70114. 70169. 70119. 70021-VWR 70022-VWR 70155. 70023-VWR 70113. 70078. 70111. 70139. 70147. 70167. 70145. 70086. 70024-VWR 70072. 70159. 70020-VWR 70090. 70112. 70053-VWR 70059-VWR 70032-VWR 70095. 70138. 70156. 70026-VWR 70173. 70115. 70025-VWR 70171. 70187. 70163. 70027-VWR 70028-VWR 70157. 70099. 70203. 70077. 70076. 70120. 70160. 70176. 70136. 70142. 70070-VWR 70192. 70089. 70080. 70209. 70082. 70151. 70052-VWR 70101. 70029-VWR 70030-VWR 70188. 70085. 70060-VWR 70031-VWR 70148. 70102. 70096. 70103. 70104. 70074. 70164. 70189. 70165. 70094. 70162. 70134. 70190. 70083. 70108. 70033. 70034-VWR 70109. 70110. 70035-VWR 70036-VWR 70154. 70132. 70084. 70037-VWR 70054-VWR 70121. 70044-VWR 70039-VWR 70137. 70041-VWR 70040-VWR 70043-VWR 70182. 70202. 70045-VWR 70126. 70153. 70133. 70149. 70046-VWR 70127. 70061-VWR 70062-VWR 70047-VWR 70048-VWR 70186. 70064-VWR 70144. 70056-VWR 70168. 70050-VWR
CultiControl freeze-dried microorganism strains derived from NCTC® cultures
Mikrobiologiske testsystemer Test for mikroorganismer
Lyophilised reference stock culture preparations containing a single strain of a microorganism for quality control of culture media, education and proficiency programs. Lyophilisation is a method for long-term preservation of microorganisms. The microorganism preparations are derived from NCTC® (National Collection of Type Cultures by the UK Health Security Agency - UKHSA).
  • One single passage from the reference NCTC® culture
  • Two years shelf life
  • More than 100 freeze-dried microorganisms available
Emballering: CultiControl microorganisms are packaged in a resealable vial that contains five (5) lyophilised pellets of a single microorganism strain, and a desiccant to prevent adverse accumulations of moisture.
Advarsel: These products are for in-vitro use only.