VWR® PerfectBlue Mini L Revolution, Gel System

Leverandør: VWR Collection
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

700-0799EA 9330 DKK
700-0799
VWR® PerfectBlue Mini L Revolution, Gel System
Elektroforesesystemer Horisontale Gelelektroforesesystemer
The 'Revolution' series includes a buffer circulation system, powered by the electrodes themselves. Hydrogen bubbles produced at the cathode pass up a diagonally positioned pipe connected to the two sides of the tank producing a gentle circulation of the buffer. The result is no pH and ionic gradient – without need of mechanical pumps.

*Based on an agarose gel of approximately 5 mm thickness

Bestillingsoplysninger: System is supplied with 2 combs (12 and 20 teeth, 1,5 mm thickness) as standard.
Bestil nu

Specifikationer

Model Mini L Revolution
Environmentally Preferable
Gelstørrelse B×L 120×140 mm
Prøvekapacitet 8 – 100
Gelvolumen 85 ml
Buffervolumen ~800 ml
Run time ≥60 h
Enhed B×L×H 170×250×95 mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR