Acetone 99,9, TraceSELECT™ Trace_analysis_inorganic, Riedel-de Haën™

Leverandør: Honeywell Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Propanon

69508-1L
69508-1LEA 798 DKK
69508-1L
Acetone 99,9, TraceSELECT™ Trace_analysis_inorganic, Riedel-de Haën™
Acetone

High purity solvents for speciation analysis by LC-ICP-MS. Rigorous purification procedures followed by UV spectroscopy, IC, and ICP-MS testing to assure high chemical purity and high UV transmittance. The blank values for metal traces in these solvents are in the ppb range or lower.

Formel: CH₃COCH₃
Molekylvægt: 58.08 g/mol
Kogepunkt: 14.3 %(V)
Smeltepunkt: –95 °C
Massefylde: 0.79 g/cm³ (20 °C)
Flammepunkt: –17 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008765
CAS-nummer: 67-64-1
EINECS: 200-662-2
UN nummer: 1090
ADR: 3 II
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Assay (GC) Min. 99.9 %
Water (Karl Fischer) Max. 0.20 %
Silver (Ag) Max. 0.5 ppb
Aluminium (Al) Max. 1 ppb
Arsenic (As) Max. 1 ppb
Gold (Au) Max. 0.5 ppb
Barium (Ba) Max. 0.5 ppb
Beryllium (Be) Max. 0.5 ppb
Bismuth (Bi) Max. 0.5 ppb
Calcium (Ca) Max. 20 ppb
Cadmium (Cd) Max. 0.5 ppb
Cerium (Ce) Max. 0.5 ppb
Cobalt (Co) Max. 0.5 ppb
Chromium (Cr) Max. 1 ppb
Cesium (Cs) Max. 0.5 ppb
Copper (Cu) Max. 1 ppb
Iron (Fe) Max. 5 ppb
Gallium (Ga) Max. 0.5 ppb
Germanium (Ge) Max. 0.5 ppb
Indium (In) Max. 0.5 ppb
Iridium (Ir) Max. 0.5 ppb
Potassium (K) Max. 10 ppb
Lithium (Li) Max. 0.5 ppb
Magnesium (Mg) Max. 1 ppb
Manganese (Mn) Max. 0.5 ppb
Molybdenum (Mo) Max. 0.5 ppb
Sodium (Na) Max. 20 ppb
Nickel (Ni) Max. 1 ppb
Lead (Pb) Max. 0.5 ppb
Palladium (Pd) Max. 0.5 ppb
Platinum (Pt) Max. 0.5 ppb
Rubidium (Rb) Max. 0.5 ppb
Rhodium (Rh) Max. 0.5 ppb
Ruthenium (Ru) Max. 0.5 ppb
Antimony (Sb) Max. 0.5 ppb
Selenium (Se) Max. 1 ppb
Tin (Sn) Max. 0.5 ppb
Strontium (Sr) Max. 0.5 ppb
Titanium (Ti) Max. 1 ppb
Thallium (Tl) Max. 0.5 ppb
Vanadium (V) Max. 0.5 ppb
Zinc (Zn) Max. 5 ppb
Zirconium (Zr) Max. 0.5 ppb
Bromide (Br) Max. 10 ppb
Chloride (Cl) Max. 25 ppb
Fluoride (F) Max. 10 ppb
Iodide (I) Max. 10 ppb
Nitrite (NO₂) Max. 10 ppb
Nitrate (NO₃) Max. 25 ppb
Phosphate (PO₄) Max. 10 ppb
Sulfate (SO₄) Max. 50 ppb
Aldehydes (as HCHO) Max. 10 ppm
Methanol (GC) Max. 0.05 %
2-propanol (GC) Max. 0.05 %
Vapor Density 2 (vs air)
Refractive Index n20/D 1.359 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR