Organic reference standard, Orange II sodium salt, analytical standard, Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
69143-25MG
69143-25MGEA 1040 DKK
69143-25MG
Organic reference standard, Orange II sodium salt, analytical standard, Supelco®
Standarder Økologiske standarder
Orange II sodium salt can undergo oxidation by hydrogen peroxide, which is catalysed by ferric ion complexed to tetra amido macrocyclic ligands(FeIII–TAML activators) to give out carbon dioxide, carbon monoxide, small aliphatic carboxylic acids and phthalic acid as the major products.
Emballering: Bottomless glass bottle. Contents are inside inserted fused cone.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d550453e0c14270fb7e889ec8117b515298cffc&vid=4d550453e0c14270fb7e889ec8117b515298cffc&prid=web&clid=SAPDAM