Multi-parameter meters, handheld, HQ series
Leverandør: Hach
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
LEV015984300A LEV0159843002 LEV0159843001 LEV015984200A LEV0159842006 LEV0159842005 LEV0159842003 LEV0159842002 LEV0159842001 LEV015984100A LEV0159841005 LEV0159841004 LEV0159841003 LEV0159841002 LEV0159841001 LEV015982200A LEV0159822009 LEV0159822008 LEV0159822006 LEV0159822005 LEV0159822004 LEV0159822003 LEV0159822002 LEV0159822001 LEV015982100A LEV0159821007 LEV0159821006 LEV0159821005 LEV0159821004 LEV0159821003 LEV0159821002 LEV0159821001 LEV015981140A LEV0159811402 LEV0159811401 LEV015981130A LEV0159811302 LEV0159811301 LEV015981110A LEV0159811104 LEV0159811103 LEV0159811102 LEV0159811101
665-0528EA 17650 DKK
665-0528 665-0529 665-0530 665-0531 665-0532 665-0533 665-0534 665-0535 665-0536 665-0537 665-0538 665-0539 665-0540 665-0541 665-0542 665-0543 665-0544 665-0545 665-0546 665-0547 665-0548 665-0549 665-0550 665-0551 665-0552 665-0553 665-0554 665-0555 665-0556 665-0557 665-0558 665-0559 663-0361 663-0362 663-0363 663-0364 663-0365 663-0366 663-0367 663-0368 663-0369 663-0370 663-0371
Multi-parameter meters, handheld, HQ series
Elektrokemiske instrumenter pH-metre
Robust and intuitive portable meters, instilling confidence in reporting and managing your results.
  • Simple calibration
  • Rugged design
  • Easy data management
  • Full range of parameters
  • Reliable quality and support
  • Lock functions include continuous/auto-stabilisation ('press to read')/at interval
The HQ series is for water quality professionals who want to perform electrochemical analysis for field and lab environments. Our new portable platform will allow you to collect intuitive, accurate measurements, manage data, and easily review results, whileMore Product Information