Orion™ Versa Star™ Pro™ Multi-Parameter Meter, Bench
Leverandør: Thermo Orion
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VSTAR00
665-0283EA 16400 DKK
665-0283
Orion™ Versa Star™ Pro™ Multi-Parameter Meter, Bench
Elektrokemiske instrumenter pH-metre
This meter offers interchangeable measurement modules that allow multiple users to customise four separate channels to meet their specific requirements.
  • Colour display with adjustable text sizes; numeric keypad; onscreen measurement stability indicator
  • Auto-read to hold a stable reading; ten password-protected methods per channel
  • Audible alarms automatically notify users when a set offset value, high/low limit value, and calibration time interval are reached
  • Uniformly mix solutions using up to two meter-controlled stirrer probes that are easy to position