pH electrodes for special laboratory applications
Leverandør: HAMILTON BONADUZ
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
238060 238100 238140 238150 238197 242064 238401
662-9206EA 2120 DKK
662-9206 662-9209 662-9211 662-9213 620-4928 662-9231 662-9232
pH electrodes for special laboratory applications
Elektroder pH-elektroder
Electrodes for special laboratory applications such as microbiological samples, serum, flat surfaces, small volumes, and samples containing fats and proteins.
  • Long-term stable Everef system
  • Different tip designs for different applications
  • Polisolve electrolyte: maintenance-free
  • 3M KCl and Protelyte electrolytes: Refillable
pH-område: 0 - 14
Bestillingsoplysninger: Supplied with individual certificate and serial number. Electrodes with S7 connector head are supplied without cable which needs to be ordered separately.