Buffer solutions
Leverandør: SI Analytics
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
285138032 285138049 285138057 285138192 285138213 285138221 285138402 285138238 285137677 285137977 285137985 285138008 285138016 285138024 285138205 285138246 285138254 285138262
SCOI285138032EA 352 DKK
SCOI285138032 662-6175 SCOI285138057 662-6180 SCOI285138213 SCOI285138221 662-1608 662-1429 SCOI285137677 SCOI285137977 662-1428 662-6181 662-6182 662-6183 SCOI285138205 662-0901 662-0902 662-0903
Buffer solutions
Buffere pH buffere
Buffer solutions intended for extremely precise calibration.
  • Highest measurement reliability
  • No preservative agents
  • Hermetically sealed in the glass ampoule
  • Sterilised with hot steam
Certificeringer: According to DIN 19 266.