Conductivity meter, SevenDirect SD30
Leverandør: Mettler - Toledo
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
30671551 30671562 30671563
662-2391EA 17050 DKK
662-2391 662-2408 662-2410
Conductivity meter, SevenDirect SD30
Elektrokemiske instrumenter Ledningsevnemålere
Robust bench top meter for accurate conductivity determinations in the lab and near production.
  • Large colour touch screen provides direct measurement and easy data entry
  • Instructions guide users through the sensor calibration process
  • Stores data, such as time stamp, sample and sensor ID with every measurement and prints or exports records automatically
  • Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP 54 in-use)
The large colour touch screen offers a direct menu access,More Product Information