Pocket pH/°C testers, Thermo Scientific™ Eutech™ Elite pH
Leverandør: EUTECH
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ELITEPH ELITEPHKIT ELITEPHSPEAR ELITEPHSPEARKI
662-2303EA 1510 DKK
662-2303 662-2280 662-2304 662-2282
Pocket pH/°C testers, Thermo Scientific™ Eutech™ Elite pH
Elektrokemiske instrumenter pH-metre
These rugged and user-friendly testers feature a simultaneous display of pH and temperature and provide accurate measurements for water analysis. Available in a standard version or with a spear tip open pore sensor for direct measurements of solid or semi-solid samples such as cheese, meat, fruit, bread and more.
  • Intuitive keypad and large graphic LCD with backlight
  • Auto-hold readings; one-push calibration
  • Easy to replace sensor
  • Lanyard provision for ‘on the go’ measurements
  • IP 67 waterproof rating
  • Standard version only: Leakproof cap with fill-line