Pocket pH and ORP testers, Pocket Pro™ / Pocket Pro™+
Leverandør: Hach
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9531100 9531000 9532000 9532100
662-1879EA 2660 DKK
662-1879 662-1878 662-2149 662-2150
Pocket pH and ORP testers, Pocket Pro™ / Pocket Pro™+
Elektrokemiske instrumenter pH-metre
These testers with integrated or replaceable sensor measure pH or ORP in a broad range of water applications. Ideal for field measurements in beverages, industrial water, wasterwater and swimming pool water.
  • pH testers with pH glass sensing element
  • Easy calibration steps and built-in diagnostics for pH testers
  • Easy-to-replace AAA batteries and a convenient sample cup make maintenance and measurements simple
  • Large LCD screen is easy to read
  • Intuitive user interface
  • Pocket Pro™+ models feature a replaceable sensor and backlit display
  • One year warranty on