VWR®, Bench Top Refractometers with Peltier Temperature Control, PLR1/PLR2
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
635-0844EA 76690 DKK
635-0844 635-0845
VWR®, Bench Top Refractometers with Peltier Temperature Control, PLR1/PLR2
Refraktometre Refraktometre, bord
Simple to use, precise bench top refractometers for use in laboratory or production environments.
  • Wide measuring range
  • Brix, RI, user scales
  • Peltier temperature control
  • 7" touch screen
  • Multilingual display
  • Intuitive menu structure
  • Store up to 4000 readings
Two distinct high precision bench top refractometers that offer users a choice based on measurement range and application type. Both models feature refractive index (nD) and sugar (Brix) scales and it is possible to load up to 25 commonly used scales from the instrument’s library.
The PLR1 isMore Product Information