Digital refractometer, Palette
Leverandør: Atago
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3480
635-0625EA 5700 DKK
635-0625
Digital refractometer, Palette
Refraktometre Håndholdte refraktometre
Digital refractometer for refractive index concentration measurement in pharmaceutical and chemical products.
  • Calibration with water
  • Simple two-button operation
  • External light interference (ELI): Accurate measurements in daylight or near a window
Leveringsinformation: Supplied with a 006p nine volt dry battery.