Slide holder, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
H445570000
631-9109EA 1400 DKK
631-9109
Slide holder, SP Bel-Art
Objektglas holdere
Polystyrene holder for up to 12 microscope slides designed for preparation or staining.
  • Inclined ribs for holding slides on their ends
  • Robust and durable white plastic design
  • Safe storage with visible hinged cases
Emballering: Sold by six items.