Silane adhesive slides, KF Frost
Leverandør: KLINIPATH
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
631-1238EA 2710 DKK
631-1238
Silane adhesive slides, KF Frost
Objektglas Adhæsionsobjektglas
These silane adhesive slides are specially designed for thermal transfer printers.
Certificeringer: CE certified according to IVDR (2017/746).