Koldlyskilder, KL 1600 LED og KL 2500 LED
Leverandør: SCHOTT AG Lighting and imaging
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
250400UK 250400EU 250400CH 150600 150600-SETEU
631-1032EA 9460 DKK
631-1032 631-1031 631-1030 631-1037 SCOC150600-SETEU
Koldlyskilder, KL 1600 LED og KL 2500 LED
Lyskilder Koldlyskilder
These LED cold light sources, for eco-friendly PURAVIS® light guides, are driven by a multiple LED module and provide a powerful illumination system for all modern stereo microscopes to illuminate all types of objects in industrial and life science applications.
  • Ecological lead-free fibre optics light guides for more visible and purer images
  • Filter slider with two positions - filter remains in the light source even when not in use
  • Compact housing with built-in handle and iron ballast for stability when used with gooseneck light guides
  • Long lifetime of at