VWR®, Immersion Oil
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
630-1808EA 214 DKK
630-1808
VWR®, Immersion Oil
Farvestoffer Mikroskopi og farvningsreagenser
Immersion oil is used to increase microscope resolution.
  • For immersing of the lens and specimen
  • Transparent oil with high refractive index
  • Increases the numerical aperture of an objective