Handheld thermo-hygrometer, HygroPalm HP32
Leverandør: ROTRONIC INSTRUMENTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HP32
620-2687EA 3830 DKK
620-2687
Handheld thermo-hygrometer, HygroPalm HP32
Hygrometre Digitale termo-hygrometre
The HygroPalm HP32 is a versatile, handheld instrument that is compatible with all HC2A and HC2 rotronic probes, and capable of performing spot measurements and logging of relative humidity, temperature and psychrometric parameters.
Applications: Field inspection and spot checks in HVAC, building management systems, agriculture and food industry as well as the pharmaceutical industry.
  • Relative humidity, temperature and psychrometric calculations
  • Graphical display for graphs and up to 4 values
  • Freely settable visual and audible alarms on every value
  • 64,000