VWR®, Digital Thermo-Hygrometer
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
620-1951EA 1450 DKK
620-1951
VWR®, Digital Thermo-Hygrometer
Hygrometre Digitale termo-hygrometre
This handheld instrument is designed to measure temperature and humidity in air conditioning and ambient air monitoring. It provides a quick response in EDP rooms, galleries, offices, workshops, storage rooms, private buildings and greenhouses.
PT1000 and capacitive-polymer sensor (Lר: 35×14 mm).
Bestillingsoplysninger: Hygrometer is supplied with a test certificate and does not come with an ISO calibration certificate. If ISO calibration certificate is required, it has to specified during ordering. If calibration certificate is required after delivery, the unit has to be sent backMore Product Information