Lactodensimeters
Leverandør: GERBER FUNKE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6600 6610
614-0250EA 492 DKK
614-0250 614-0251
Lactodensimeters
Flydevægte
This lactodensimeter according to Gerber is designed with a thermometer.
  • Thermometer included in stem or body
  • Standard model with negative scale
  • Calibrated temperature of 20 °C