Automatic tilt measures

Leverandør: GERBER FUNKE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

3421 3420
614-0148EA 726 DKK
614-0148 614-0147
Automatic tilt measures
Flaskedispensere
Glass, with rubber stopper.

  • Superior quality models
  • Storage bottle of 500/250 ml
  • For milk analysis applications

Colour: Clear
Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR