epT.I.P.S.® Pipette Tips, BioBased Reloads
Leverandør: EPPENDORF
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0030075.420 0030075.439 0030075.447 0030075.455 0030075.463 0030075.471
613-7278EA 490 DKK
613-7278 613-7279 613-7280 613-7281 613-7282 613-7283
epT.I.P.S.® Pipette Tips, BioBased Reloads
Pipettespidser
Eppendorf epT.I.P.S.® BioBased Reloads in 'Biopur' grade innovate with cutting edge eco-friendly design. The tips are made from 90% plant-based material in a reloadable format. They ensure precision and purity, and are ideal for sensitive applications prioritising environmental responsibility.
  • Improve the laboratory carbon footprint with epT.I.P.S.® BioBased Biopur® in the new Eppendorf Reloads
  • Produced from recycled feedstock in second generation, derived solely from waste and residue streams from vegetable oil production
  • Significantly reduce laboratory