ClipTip™ Pipette Tips
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
94410210 94410213 94410217 94410218 94410250 94410253 94410257 94410258 94410310 94410313 94410317 94410318 94410510 94410513 94410517 94410518 94410710 94410713 94410717 94410718 94410040 94410043 94410050 94410053 94410100 94410103 94410150 94410153 94410810 94410813 94410817 94410818 94410610 94410618 94410617 94410060 94410063 94410070 94410073 94410613
613-2205EA 348 DKK
613-2205 613-2206 613-2207 613-2208 613-2209 613-2210 613-2211 613-2212 613-2213 613-2214 613-2215 613-2216 613-2217 613-2218 613-2219 613-2220 613-2222 613-2223 613-2224 613-2225 613-6031 613-6032 613-6025 613-6026 613-6027 613-6028 613-6029 613-6030 613-6033 613-6034 613-6035 613-6036 613-6194 613-7447 613-7446 613-6861 613-6862 613-6192 613-6193 613-6195
ClipTip™ Pipette Tips
Pipettespidser
ClipTip™ pipette tips provide security with a unique and innovative interlocking technology that ensures a complete seal on every channel with minimal tip attachment and ejection force.
  • Pipette tips feature 'clips' that lock tips in place and forms a complete seal
  • Every ClipTip™ pipette tip utilises low retention technology to enable maximium sample recovery
  • Certified free of RNase, DNase, DNA, ATP and endotoxin contamination
  • Each tip insert has ink-jetted lot and volume information; the convenient colour key makes it easy to match the colour coding of the