Enkeltkanalpipetter, mekaniske, variabel volumen, mLINE®
Leverandør: Sartorius
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
725080 725070 725060 725050 725020 725030 725010 725090
613-4093EA 3200 DKK
613-4093 613-4092 613-4091 613-4090 613-4089 613-5078 613-5125 613-5147
Enkeltkanalpipetter, mekaniske, variabel volumen, mLINE®
Pipetter Automatiske pipetter
mLINE® pipettes offer a unique combination of new features for higher accuracy and precision. They include built-in replaceable tip cone filters, which prevent pipette contamination and damage.
  • Clear and easy to read display with big numbers and small increments
  • Equipped with special spring-loaded tip cones (i.e. the Optiload mechanism) allowing tip loading and ejection using minimum force
  • Plunger technology results in the lowest pipetting forces
  • Volume range colour coding for easy pipette selection, five colour coding caps
  • Thermal insulation of the