Filter tips, SafetySpace™
Leverandør: Sartorius
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
783201 790011F 790021F 790101F 790201F 790301F 791001F 791211F LH-795001F LH-XF780201
613-5091EA 1480 DKK
613-5091 613-0857 613-0858 613-0859 613-0860 613-0898 613-0899 613-0900 613-6746 BHITLH-XF780201
Filter tips, SafetySpace™
Pipettespidser
These filter tips have more space between the sample and the filter than conventional filter tips. This virtually eliminates the risk of an expensive or a contagious sample permeating the filter.
  • PP with PE filter, sterile, non autoclavable
  • Higher filter position prevents liquid contacting the filter reducing the risk of pipette or sample contamination
  • Electron beam sterilisation
  • Certified as DNase-, RNase- and endotoxin-free (certificates can be downloaded on supplier website)
Ideal for molecular biology, microbiology and cell culture applications,More Product Information