Pipette tips, universal, Corning®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4803 4804 4806 4844 4845 4846 4860 4862 4863 4864 4865 4866 4867 4868 9032 4713
613-4239EA 2730 DKK
613-4239 613-4240 613-4241 613-4246 613-4247 613-4248 613-4249 613-4997 613-4998 613-4999 613-4250 613-5000 613-5001 612-5007 613-5003 613-5500
Pipette tips, universal, Corning®
Pipettespidser
Corning® universal fit tips are designed to provide a reliable fit with all major brand pipettors.
  • Beveled orifice helps ensure accurate fluid delivery
  • All tips are certified RNase-/DNase-free and non pyrogenic
  • Stack rack tips feature a stack of five racks, each containing 96 tips, for a total of 480 tips in a space-saving design
  • Bulk packed tips are non sterile and very economical
1 to 200 µl universal fit tips are graduated at the 10 µl, 50 µl, and 100 µl volumes.
Bestil nu
Specifikationer
Environmentally Preferable Lav fremstillingspåvirkning Lav fremstillingspåvirkning
Miljøcertificering I-RECs, ISO 14001 - Reynosa I-RECs, ISO 14001 - Monterrey