VWR® Traceable® Calibrated Timer
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
609-0195EA 558 DKK
609-0195
VWR® Traceable® Calibrated Timer
Alarmure Digitale timere
Personalise with coloured faceplates.
  • Unique memory returns channel to previously programmed setting at touch of a key
  • Four independent timing channels
  • NIST-traceable
  • Counts up during alarming indicating elapsed time since alarming, 12/24 hour clock
  • 1 year warranty
Four independent timing channels feature unique audible alarms for simultaneous timing. Count up or down from 100 hours for longer duration experiments. Memory function returns the display to the programmed time.
Traceable® Products have an individually numbered Traceable® CertificateMore Product Information