Mouse IgG2b Isotype Control [clone: MPC-11] (PE (Phycoerythrin)-Cy7®)
Leverandør: Tonbo Biosciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
60-4732-U100
60-4732-U100EA 1050 DKK
60-4732-U100
Mouse IgG2b Isotype Control [clone: MPC-11] (PE (Phycoerythrin)-Cy7®)
Antistoffer
The MPC-11 immunoglobulin is useful as an isotype-matched control. The MPC-11 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2b antibodies.