Anti-PTPRC Mouse antibody (PE (Phycoerythrin)-Cy7®) [clone: HI30]
Leverandør: Tonbo Biosciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
60-0459-T025 60-0459-T100
60-0459-T025EA 652 DKK
60-0459-T025 60-0459-T100
Anti-PTPRC Mouse antibody (PE (Phycoerythrin)-Cy7®) [clone: HI30]
Antistoffer
The HI30 antibody reacts with human CD45, one of the most abundant hematopoietic markers and one that is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). CD45 is a protein tyrosine phosphatase existing in several isoforms, each being generated and expressed in cell-specific patterns. With its broad cell distribution, CD45 is critical for many leukocyte functions, regulating signal transduction and cell activation associated with the T cell receptor, B cell receptor, and IL-2 receptor. Other forms of CD45, with restricted cellular expression, include CD45R (B220), CD45RA, CD45More Product Information