Supelco® Organic Reference Standard, Urea

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

56180-1G
56180-1GEA 394 DKK
56180-1G
Supelco® Organic Reference Standard, Urea
Standarder Økologiske standarder
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa37b91cff89d8a20d47941ad62044595432a0ed&vid=aa37b91cff89d8a20d47941ad62044595432a0ed&prid=web&clid=SAPDAM

Bestil nu

Specifikationer

Synonym(s) Carbamide; Carbonyldiamide
Environmentally Preferable
M-Clarity quality level MQ100
Densitet 1,335 g/ml at 25 °C (lit.)
CAS-nr 57-13-6
MDL No. MFCD00008022
Formel NH₂CONH₂
Molecular weight 60,06
Melting point 132 - 135 °C (lit.)
Shipping conditions STD
Storage conditions RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR