J.T.Baker® BAKER ANALYZED®, Buffer Solutions
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7261.1000EA 440 DKK
7261.1000 7262.1000 7267.1000 7263.1000 7265.1000 7268.1000 7270.1000 5678.1000 5677.1000 5643.1000 5609.0500 5609.1000 5608.9010 5608.5000PE 5608.2500PE 5608.1000 5608.0500 5606.9010 5606.5000PE 5606.2500PE 5606.1000 5606.0500 5609.9010
J.T.Baker® BAKER ANALYZED®, Buffer Solutions
Buffere pH buffere
Ready-to-use pH buffers are manufactured in large lots to stringent specifications and utilise quality control procedures to reduce lot-to-lot variability. You can eliminate the time and expense of preparation and standardisation of solutions, helping streamline high-volume analytical work in applications such as quality control.
Contain 0,01% Sodium benzoate.
Certificeringer: This buffer is traceable to NIST Standard reference buffer.