Capillary columns, TraceGOLD™ TG-200MS Fast
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
26084-2760
555-8891EA 6140 DKK
555-8891
Capillary columns, TraceGOLD™ TG-200MS Fast
Kromatografikolonner
Exceptionally inert mid-polarity columns with selectivity and elution order optimised for difficult separations.
  • Low bleed, polar phase, trifluoropropyl methylpolysiloxane
  • High thermal stability - maximum temperatures up to 320/340 °C
  • Suitable for use with sensitive detectors including ECD, NPD and MS