Coated polysaccharide columns, normal phase, CHIRALPAK®
Leverandør: CHIRAL TECHNOLOGIES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
19045 19094 20035 20045 20094 20325 20324 19032 19042 19335 19345 20032 20042 20335 20345 19394 20394 19025 19035 20025 19324 19325 19311 19337 20311 20337 19435 19023 19524 19525 19511 47325 20511 20594 20525 20524 20522 19594 19523 19522 20523 61394 47324 47524 47594 47525 47311 47522 47511 47394 47345 47337 47335 19224 19225 19226 19322 19323 19423 20224 20225 20226 20322 20323 20423 47224 47225 47226 47322 47323 47423 47435 47445 61224 61225 61226 61322 61323 61335 61337 61345 61423 61435 61445 47595 20595 19595 47S84 61S84 61S82 20S84 47S82 20S83 19S82 61S83 19S83 19S84 20S82 47S83 19G22 19G24 19G25 19G94 19G35 19G45 20G22 20G24 20G94 20G35 20G45 47G22 47G24 47G25 47G94 47G35 47G45 61G22 61G24 61G25 61G94 61G35 61G45
554-0060EA 110420 DKK
554-0060 554-0062 554-0076 554-0078 554-0080 554-0085 554-0084 554-0057 554-0059 554-0069 554-0070 554-0075 554-0077 554-0087 554-0088 554-0121 554-0126 554-0056 554-0058 554-0074 554-0066 554-0067 554-0065 554-5709 554-3759 554-3758 554-3767 554-4068 554-2461 554-2462 554-2458 554-5738 554-2464 554-2469 554-2468 554-2467 554-2465 554-2463 554-2460 554-2459 554-2466 554-3413 554-5737 554-5745 554-5747 554-5746 554-5736 554-5744 554-5743 554-5742 554-5741 554-5740 554-5739 554-4067 554-4066 554-4065 554-4064 554-4063 554-4071 554-4072 554-4073 554-4074 554-4075 554-4076 554-4077 554-3397 554-3398 554-3399 554-3400 554-3401 554-3402 554-3403 554-3404 554-3405 554-3406 554-3407 554-3408 554-3409 554-3410 554-3411 554-3412 554-3414 554-3415 554-3416 554-5748 554-5719 554-5710 554-4592 554-4601 554-4599 554-4586 554-4590 554-4585 554-4581 554-4600 554-4582 554-4583 554-4584 554-4591 DAIC19G22 DAIC19G24 DAIC19G25 DAIC19G94 DAIC19G35 DAIC19G45 DAIC20G22 DAIC20G24 DAIC20G94 DAIC20G35 DAIC20G45 DAIC47G22 DAIC47G24 DAIC47G25 DAIC47G94 DAIC47G35 DAIC47G45 DAIC61G22 DAIC61G24 DAIC61G25 DAIC61G94 DAIC61G35 DAIC61G45
Coated polysaccharide columns, normal phase, CHIRALPAK®
Kromatografikolonner
These columns are packed with Daicel’s coated polysaccharide-based chiral stationary phases (CSPs) which are manufactured by physically coating the polysaccharide derivatives onto silica matrices. Due to this manufacturing process, a choice of mobile phase components is typically narrower than with immobilised columns in order to minimise damage to the CSPs.