Coated polysaccharide columns, reversed phase
Leverandør: CHIRAL TECHNOLOGIES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
14625 14724 20724 19724 14711 17711 17724 19711 20711 17794 19824 19811 14822 19822 20824 14824 17822 17823 14823 14811 20823 20822 20811 17811 19894 19823 17824 61824 61823 61822 61811 47824 42824 42823 42822 42811 47823 47822 47811 42711 63811 63822 63823 63824 19892 19893 19895 14825
554-0031EA 21860 DKK
554-0031 554-0033 554-0090 554-0072 554-3743 554-0048 554-3746 554-3768 554-3757 554-3747 554-5714 554-5711 DAIC14822 554-5712 DAIC20824 DAIC14824 DAIC17822 DAIC17823 DAIC14823 DAIC14811 DAIC20823 DAIC20822 DAIC20811 DAIC17811 554-5717 554-5713 DAIC17824 DAIC61824 DAIC61823 DAIC61822 DAIC61811 DAIC47824 DAIC42824 DAIC42823 DAIC42822 DAIC42811 DAIC47823 DAIC47822 DAIC47811 554-3396 DAIC63811 DAIC63822 DAIC63823 DAIC63824 554-5715 554-5716 554-5718 DAIC14825
Coated polysaccharide columns, reversed phase
Kromatografikolonner
These columns have the same coated chiral selector as found in the normal phase stationary phases, however in comparison, they use a different bounded silica as the base material which stabilises the CSP.
Reversed phase analytical columns were developed specifically for aqueous-organic mobile phases. They are suited for applications where the sample is presented in aqueous media (e.g. biological samples) or for samples that require flexibility in term of pH range.
Extreme pH values must be avoided as they can damage the silica gel used in these columns. These columns are alsoMore Product Information