Spray bottle
Leverandør: witeg Labortechnik
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2.147.000.C
552-6001EA 919 DKK
552-6001
Spray bottle
Forstøvere
Glass spray bottle with rubber bulb for simple manual operation of the solution spray.
  • Connection for compressed air generator
100 ml capacity