VWR®, Storage Boxes for Sample Vials and Bottles, PP
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
548-1995AEA 222 DKK
548-1995A 548-1996A 548-1997A 548-1998A 548-1999A 548-2000A 548-1511A 548-1443A 548-1445A 548-1444A 548-1447A 548-1448A 548-1446A 548-1512A 548-1513A
VWR®, Storage Boxes for Sample Vials and Bottles, PP
Bokse Kasser til Vials
A broad variety of PP storage boxes to securely hold a wide range of vial sizes and bottles. Perfect for space-saving storage in the laboratory or in fridges.
  • Transparent lid
  • Chemically resistant
  • Stackable and robust
  • Temperature resistant from –80 to 100 °C
  • Easy sample identification (alphanumeric coding) for boxes with 81 cavities
Transperent lids prevents condensation on the closures and thus avoid contamination in the refrigerator.