VWR® ND8, krympehalshætteglas
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
548-0035AEA 1520 DKK
548-0035A 548-0036A 548-0037A 548-0078A 548-0080A 548-0339A 548-0340A 548-0341A 548-0783A 548-0831A
VWR® ND8, krympehalshætteglas
Hætteglas
These vials are for use on instruments from the following manufacturers: Agilent, Beckman, Carlo Erba, CTC, Fisons, PerkinElmer, Shimadzu, Thermo Scientific, VWR®/Hitachi, more.
  • Flat, round or conical bottom
  • In clear or amber glass
  • For almost all autosamplers
Broad selection of crimp neck vials ND8 available: Crimp neck vials and micro-vials ND8 can be closed with 8 mm aluminium caps, 9 mm PE caps or with 8 mm push-on caps. However, micro-vials often need an adapter to run in the autosampler. They often have a conical bottom shape, so need an adapter to stand.