VWR® Digital Rotary Evaporator, LCD
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
531-1365EA 26040 DKK
531-1365 531-1366 531-1367 531-1368 531-1377
VWR® Digital Rotary Evaporator, LCD
Fordampere Rotationsfordampere
This is an essential instrument for use in chemical laboratories for efficient and gentle removal of solvents from samples by evaporation. It offers an excellent distilling solution in a wide range of applications.
  • Digital display shows both rotation speed and heating temperature allowing for optimal control of all distillation processes
  • Automatic motor lift releases the evaporating flask to a safe position in case of power failure
  • 5 L heating bath with wide temperature range (ambient to 180 °C); water/oil heating mode can only be changed via a switch