Recirculating chillers, F-100 / F-108 / F-114 / F-125
Leverandør: BUCHI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
11056460
531-0858EA 26730 DKK
531-0858
Recirculating chillers, F-100 / F-108 / F-114 / F-125
Cirkulatorsystemer Lukkede cirkulatorsystemer Køletermostatbade
The F-series chillers provide the final step to a fully integrated evaporation or distillation process, leading to several key benefits:
  • Up to 75% less energy consumption
  • Reduced heat emission
  • Easy handling and mobility
  • Optimised distillation capacity
  • Increased process safety due to communication of all connected instruments
F-100: Cost effective, small recirculating chiller that fulfils the basic cooling requirements. The perfect replacement of tap water chilling with small distillation instruments. It cools to a fixed temperature of 10 °C and isMore Product Information