Conical Tubes with Screw Caps
Leverandør: EPPENDORF
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0030122.178 0030122.259 0030122.267 0030122.160 0030122.186 0030122.194 0030122.224 0030122.437 0030122.500 0030122.410 0030122.429 0030122.445 0030113.489 0030113.497 0030122.526 0030122.534 0030122.542
525-1052EA 1590 DKK
525-1052 EPPE0030122.259 EPPE0030122.267 525-1051 525-1054 525-1050 525-1053 EPPE0030122.437 EPPE0030122.500 EPPE0030122.410 EPPE0030122.429 EPPE0030122.445 EPPE0030113.489 EPPE0030113.497 525-1612 525-1613 525-1614
Conical Tubes with Screw Caps
Rør Centrifugerør
Eppendorf® conical tubes with screw caps offer reliable sample containment and leak-free storage. SafeCode variants enhance sample identification, while the BioBased options, manufactured using renewable resources, provide an eco-friendly alternative without compromising performance.
  • Polypropylene (PP) tubes, High-Density Polyethylene (HDPE) screw caps
  • Ensures leak-free storage and protects sample integrity
  • Suited for a wide range of laboratory tasks, enhancing flexibility
  • High-quality materials for long-lasting, robust performance
  • Clear, visible