Eppendorf® DNA LoBind® Microcentrifuge Tubes
Leverandør: EPPENDORF
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0030108.078 0030108.035 0030108.051 0030108.310 0030122.208 0030122.232 0030122.348 0030108.400 0030108.418 0030108.426 0030108.523 0030122.275
525-0131EA 150 DKK
525-0131 525-0129 525-0130 525-0793 525-0985 525-0986 EPPE0030122.348 EPPE0030108.400 EPPE0030108.418 EPPE0030108.426 525-1602 EPPE0030122.275
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Rør Mikrocentrifugerør
Unleash the power of precision in DNA research with Eppendorf® DNA LoBind® tubes. Engineered with proprietary LoBind® technology, these microcentrifuge tubes redefine sample handling, minimising the risk of loss and maximising DNA recovery. Ideal for low-concentration applications, including PCR, qPCR, and DNA sequencing, these tubes ensure accuracy, purity, and optimal yields.
  • Surface-coating-free, including substances like silicone, to reduce the risk of sample interference
  • Lot-certified PCR clean purity grade guarantees freedom from human DNA, DNase,
More Product Information
Bestil nu
Specifikationer
Environmentally Preferable Lav fremstillingspåvirkning
Miljøcertificering Renewable Energy Certificate, ISO 14001